tert-Butyl 4-bromo-3,5-dimethyl-1H-pyrazole-1-carboxylate ≥97%

Supplier: AOBChem USA

Warning

57311-25G 57311-5G
76943-366EA 630.18 USD
76943-366 76943-368
tert-Butyl 4-bromo-3,5-dimethyl-1H-pyrazole-1-carboxylate ≥97%
tert-Butyl 4-bromo-3,5-dimethyl-1H-pyrazole-1-carboxylate
Formula: C₁₀H₁₅BrN₂O₂
MW: 275.142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12756451
CAS Number: 1040276-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR