Methyl 2-Isopropylbenzoate ≥95%

Supplier: AOBChem USA

Warning

57154-1G 57154-25G 57154-500MG 57154-5G
76943-350EA 206.88 USD
76943-350 76943-352 76943-354 76943-356
Methyl 2-Isopropylbenzoate ≥95%
Methyl 2-Isopropylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD20484275
CAS Number: 6623-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR