5-Iodo-1-phenyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

56559-1G 56559-25G 56559-500MG 56559-5G
76943-326EA 233.4 USD
76943-326 76943-328 76943-330 76943-332
5-Iodo-1-phenyl-1H-pyrazole ≥97%
5-Iodo-1-phenyl-1H-pyrazole
Formula: C₉H₇IN₂
Storage Temperature: Refrigerator
MDL Number: MFCD27944668
CAS Number: 141998-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR