Methyl 3-chloro-2-fluoro-6-iodobenzoate ≥97%

Supplier: AOBChem USA

Warning

56318-1G 56318-25G 56318-500MG 56318-5G
76943-312EA 194.15 USD
76943-312 76943-314 76943-316 76943-318
Methyl 3-chloro-2-fluoro-6-iodobenzoate ≥97%
Methyl 3-chloro-2-fluoro-6-iodobenzoate
Formula: C₈H₅ClFIO₂
MW: 314.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34530511
CAS Number: 2091029-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR