5-Iodo-1-methyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Iodo-1-methylpyrazole

55975-25G 55975-5G
76943-300EA 254.62 USD
76943-300 76943-302
5-Iodo-1-methyl-1H-pyrazole ≥97%
5-Iodo-1-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₅IN₂
MW: 208.00 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11109327
CAS Number: 34091-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR