2,4,5-Trichlorothioanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl(2,4,5-trichlorophenyl)sulphane

54813-25G 54813-5G
76943-188EA 198.39 USD
76943-188 76943-190
2,4,5-Trichlorothioanisole ≥97%
2,4,5-Trichlorothioanisole

Precaución: for Research Use Only

Formula: C₇H₅Cl₃S
Storage Temperature: Ambient
MDL Number: MFCD00044426
CAS Number: 4163-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR