Methyl benzothiophene-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl benzo[b]thiophene-3-carboxylate

54359-1G 54359-25G 54359-500MG 54359-5G
76943-164EA 155.96 USD
76943-164 76943-166 76943-168 76943-170
Methyl benzothiophene-3-carboxylate ≥97%
Methyl benzothiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈O₂S
MW: 192.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203855
CAS Number: 22913-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR