Methyl 2-bromo-4-fluoro-5-methylbenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
53546-25G
53546-5G
76943-118EA
595.17
USD
76943-118
76943-120
Methyl 2-bromo-4-fluoro-5-methylbenzoate ≥97%
Methyl 2-bromo-4-fluoro-5-methylbenzoate
Formula:
C₉H₈BrFO₂ Storage Temperature: Ambient |
CAS Number:
1437780-13-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...