6-Hydroxy-5-iodopyridine-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

53320-25G 53320-5G
76943-106EA 537.88 USD
76943-106 76943-108
6-Hydroxy-5-iodopyridine-3-carboxylic acid ≥97%
6-Hydroxy-5-iodopyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₄INO₃
Storage Temperature: Ambient
MDL Number: MFCD24039043
CAS Number: 365413-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR