Methyl 5-formyl-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

53243-1G 53243-25G 53243-500MG 53243-5G
76943-090EA 215.37 USD
76943-090 76943-092 76943-094 76943-096
Methyl 5-formyl-2-methylbenzoate ≥97%
Methyl 5-formyl-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MW: 178.185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21099506
CAS Number: 675148-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR