6-Chloro-2-fluoro-m-toluic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-2-fluoro-3-methylbenzoic acid

52859-25G 52859-5G
76943-078EA 262.05 USD
76943-078 76943-080
6-Chloro-2-fluoro-m-toluic acid ≥97%
6-Chloro-2-fluoro-m-toluic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631369
CAS Number: 32890-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR