6-Formylnicotinic acid ≥97%

Supplier: AOBChem USA

Warning

52698-1G 52698-25G 52698-500MG 52698-5G
76943-062EA 292.81 USD
76943-062 76943-064 76943-066 76943-068
6-Formylnicotinic acid ≥97%
6-Formylnicotinic acid
Formula: C₇H₅NO₃
MW: 151.119 g/mol
Storage Temperature: Refrigerator
CAS Number: 6542-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR