Ethyl-2-bromonicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-bromonicotinate

52394-1G 52394-25G 52394-5G
76943-038EA 84.88 USD
76943-038 76943-040 76943-042
Ethyl-2-bromonicotinate ≥97%
Ethyl-2-bromonicotinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11506200
CAS Number: 53087-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR