Methyl 2-bromo-5-methoxyisonicotinate ≥97%

Supplier: AOBChem USA

Warning

51477-1G 51477-500MG 51477-5G
76942-980EA 589.87 USD
76942-980 76942-982 76942-984
Methyl 2-bromo-5-methoxyisonicotinate ≥97%
Methyl 2-bromo-5-methoxyisonicotinate
Formula: C₈H₈BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD18257536
CAS Number: 1256819-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR