6-Fluoro-[1,1'-biphenyl]-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

50794-1G 50794-25G 50794-500MG 50794-5G
76942-956EA 203.7 USD
76942-956 76942-958 76942-960 76942-962
6-Fluoro-[1,1'-biphenyl]-3-carboxylic acid ≥97%
6-Fluoro-[1,1'-biphenyl]-3-carboxylic acid
Formula: C₁₃H₉FO₂
MW: 216.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11577106
CAS Number: 1082484-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR