Methyl 3-bromo-2,4-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

50633-1G 50633-25G 50633-500MG 50633-5G
76942-940EA 296 USD
76942-940 76942-942 76942-944 76942-946
Methyl 3-bromo-2,4-difluorobenzoate ≥97%
Methyl 3-bromo-2,4-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09261258
CAS Number: 935534-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR