Ethyl 4-fluoro-3-hydroxybenzoate ≥95% research grade

Supplier: AOBChem USA

Warning

50031-1G 50031-25G 50031-500MG 50031-5G
76942-926EA 155.96 USD
76942-926 76942-928 76942-930 76942-932
Ethyl 4-fluoro-3-hydroxybenzoate ≥95% research grade
Ethyl 4-fluoro-3-hydroxybenzoate
Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD12025283
CAS Number: 351317-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR