Methyl 2,3-difluoro-4-hydroxybenzoate ≥95%

Supplier: AOBChem USA

Warning

49965-25G 49965-5G
76942-918EA 537.88 USD
76942-918 76942-920
Methyl 2,3-difluoro-4-hydroxybenzoate ≥95%
Methyl 2,3-difluoro-4-hydroxybenzoate
Formula: C₈H₆F₂O₃
MW: 188.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042793
CAS Number: 219685-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR