Methyl 4-bromo-2-fluoro-5-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

49604-1G 49604-25G 49604-500MG 49604-5G
76942-892EA 439.22 USD
76942-892 76942-894 76942-896 76942-898
Methyl 4-bromo-2-fluoro-5-(trifluoromethyl)benzoate ≥97%
Methyl 4-bromo-2-fluoro-5-(trifluoromethyl)benzoate
Formula: C₉H₅BrF₄O₂
MW: 301.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30742530
CAS Number: 2090828-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR