Methyl 3-bromo-5-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

49137-1G 49137-500MG 49137-5G
76942-854EA 468.92 USD
76942-854 76942-856 76942-858
Methyl 3-bromo-5-(methylthio)benzoate ≥97%
Methyl 3-bromo-5-(methylthio)benzoate
Formula: C₉H₉BrO₂S
MW: 261.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22488918
CAS Number: 453566-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR