Ethyl 2,6-difluoro-3-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

49059-1G 49059-25G 49059-500MG 49059-5G
76942-842EA 292.81 USD
76942-842 76942-844 76942-846 76942-848
Ethyl 2,6-difluoro-3-methylbenzoate ≥95%
Ethyl 2,6-difluoro-3-methylbenzoate
Formula: C₁₀H₁₀F₂O₂
MW: 200.182 g/mol
Storage Temperature: Ambient
CAS Number: 773135-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR