Ethyl 3-ethoxy-4-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

48965-1G 48965-25G 48965-500MG 48965-5G
76942-814EA 159.14 USD
76942-814 76942-816 76942-818 76942-820
Ethyl 3-ethoxy-4-methylbenzoate ≥95%
Ethyl 3-ethoxy-4-methylbenzoate
Formula: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD26402596
CAS Number: 1196047-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR