Ethyl 3,6-dichloro-2-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

48696-1G 48696-25G 48696-500MG 48696-5G
76942-798EA 439.22 USD
76942-798 76942-800 76942-802 76942-804
Ethyl 3,6-dichloro-2-fluorobenzoate ≥97%
Ethyl 3,6-dichloro-2-fluorobenzoate
Formula: C₉H₇Cl₂FO₂
MW: 237.055 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14698218
CAS Number: 1214329-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR