N-Methyl-3,5-bis(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

47719-1G 47719-25G 47719-5G
76942-702EA 79.57 USD
76942-702 76942-704 76942-706
N-Methyl-3,5-bis(trifluoromethyl)benzamide ≥97%
N-Methyl-3,5-bis(trifluoromethyl)benzamide
Formula: C₁₀H₇F₆NO
Storage Temperature: Ambient
CAS Number: 948294-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR