Methyl 5-bromo-3-chloro-2-fluorobenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
47260-1G
47260-25G
47260-500MG
47260-5G
76942-676EA
379.81
USD
76942-676
76942-678
76942-680
76942-682
Methyl 5-bromo-3-chloro-2-fluorobenzoate ≥97%
Methyl 5-bromo-3-chloro-2-fluorobenzoate
Formula:
C₈H₅BrClFO₂ MW: 267.48 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD24107276 CAS Number: 1535973-93-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...