Methyl 4-hydroxy-2-isopropylbenzoate ≥97% research grade

Supplier: AOBChem USA

Warning

46904-1G 46904-500MG 46904-5G
76942-662EA 699.14 USD
76942-662 76942-664 76942-666
Methyl 4-hydroxy-2-isopropylbenzoate ≥97% research grade
Methyl 4-hydroxy-2-isopropylbenzoate
Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
CAS Number: 1529775-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR