Methyl [1,1'-biphenyl]-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

46722-1G 46722-25G 46722-500MG 46722-5G
76942-646EA 317.21 USD
76942-646 76942-648 76942-650 76942-652
Methyl [1,1'-biphenyl]-3-carboxylate ≥97%
Methyl [1,1'-biphenyl]-3-carboxylate
Formula: C₁₄H₁₂O₂
MW: 212.244 g/mol
Storage Temperature: Ambient
CAS Number: 16606-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR