Ethyl 2-ethoxy-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

46365-1G 46365-25G 46365-500MG 46365-5G
76942-622EA 126.25 USD
76942-622 76942-624 76942-626 76942-628
Ethyl 2-ethoxy-4-methylbenzoate ≥97%
Ethyl 2-ethoxy-4-methylbenzoate
Formula: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD09701458
CAS Number: 88709-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR