Methyl 2-methoxy-[1,1'-biphenyl]-4-carboxylate ≥97%

Supplier: AOBChem USA

Warning

46308-1G 46308-25G 46308-500MG 46308-5G
76942-606EA 332.07 USD
76942-606 76942-608 76942-610 76942-612
Methyl 2-methoxy-[1,1'-biphenyl]-4-carboxylate ≥97%
Methyl 2-methoxy-[1,1'-biphenyl]-4-carboxylate
Formula: C₁₅H₁₄O₃
Storage Temperature: Ambient
CAS Number: 175152-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR