Methyl indole-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 1H-indole-1-carboxylate

46117-25G 46117-5G
76942-594EA 630.18 USD
76942-594 76942-596
Methyl indole-1-carboxylate ≥95%
Methyl indole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Storage Temperature: Ambient
CAS Number: 39203-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR