Methyl 6-ethoxypicolinate ≥97%

Supplier: AOBChem USA

Warning

46093-1G 46093-25G 46093-5G
76942-582EA 117.76 USD
76942-582 76942-584 76942-586
Methyl 6-ethoxypicolinate ≥97%
Methyl 6-ethoxypicolinate
Formula: C₉H₁₁NO₃
MW: 181.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17169085
CAS Number: 55980-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR