Methyl 3-bromo-5-(trifluoromethoxy)benzoate ≥97%

Supplier: AOBChem USA

Warning

46056-1G 46056-25G 46056-5G
76942-554EA 94.43 USD
76942-554 76942-556 76942-558
Methyl 3-bromo-5-(trifluoromethoxy)benzoate ≥97%
Methyl 3-bromo-5-(trifluoromethoxy)benzoate

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD09999511
CAS Number: 1306763-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR