Methyl biphenyl-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl [1,1'-biphenyl]-2-carboxylate

45983-1G 45983-25G 45983-500MG 45983-5G
76942-538EA 155.96 USD
76942-538 76942-540 76942-542 76942-544
Methyl biphenyl-2-carboxylate ≥97%
Methyl biphenyl-2-carboxylate
Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00452250
CAS Number: 16605-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR