Ethyl 4-methylthiophene-2-carboxylate ≥97%
Supplier: AOBChem USA
|
Warning
|
45950-1G
45950-25G
45950-5G
76942-524EA
84.88
USD
76942-524
76942-526
76942-528
Ethyl 4-methylthiophene-2-carboxylate ≥97%
Ethyl 4-methylthiophene-2-carboxylate
Precaución: for Research Use Only
Formula:
C₈H₁₀O₂S Storage Temperature: Refrigerator |
MDL Number:
MFCD12025317 CAS Number: 14282-79-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...