Methyl 2,4,6-trimethylbenzoate ≥95%

Supplier: AOBChem USA

Warning

45919-1G 45919-25G 45919-5G
76942-514EA 76.39 USD
76942-514 76942-516 76942-518
Methyl 2,4,6-trimethylbenzoate ≥95%
Methyl 2,4,6-trimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00184961
CAS Number: 2282-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR