Ethyl 1-methyl-1H-pyrazole-5-carboxylate ≥97%

Supplier: AOBChem USA

Warning

45895-25G 45895-5G
76942-494EA 310.85 USD
76942-494 76942-496
Ethyl 1-methyl-1H-pyrazole-5-carboxylate ≥97%
Ethyl 1-methyl-1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08700760
CAS Number: 197079-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR