Methyl 2,6-dichloro-4-methylnicotinate ≥97%

Supplier: AOBChem USA

Warning

45870-1G 45870-25G 45870-5G
76942-484EA 82.76 USD
76942-484 76942-486 76942-488
Methyl 2,6-dichloro-4-methylnicotinate ≥97%
Methyl 2,6-dichloro-4-methylnicotinate
Formula: C₈H₇Cl₂NO₂
MW: 220.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15144593
CAS Number: 1013648-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR