Methyl 5-bromo-2,4-dimethylbenzoate ≥95%

Supplier: AOBChem USA

Warning

45844-1G 45844-500MG 45844-5G
76942-470EA 670.49 USD
76942-470 76942-472 76942-474
Methyl 5-bromo-2,4-dimethylbenzoate ≥95%
Methyl 5-bromo-2,4-dimethylbenzoate
Formula: C₁₀H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD11112985
CAS Number: 152849-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR