Methyl 5-hydroxy-2-methoxybenzoate ≥95% research grade

Supplier: AOBChem USA

Warning

45668-1G 45668-25G 45668-500MG 45668-5G
76942-456EA 272.66 USD
76942-456 76942-458 76942-460 76942-462
Methyl 5-hydroxy-2-methoxybenzoate ≥95% research grade
Methyl 5-hydroxy-2-methoxybenzoate
Formula: C₉H₁₀O₄
MW: 182.173 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01940408
CAS Number: 87513-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR