7-Bromo-1H-pyrazolo[4,3-c]pyridine ≥97%

Supplier: AOBChem USA

Warning

45584-1G 45584-25G 45584-500MG 45584-5G
76942-434EA 292.81 USD
76942-434 76942-436 76942-438 76942-440
7-Bromo-1H-pyrazolo[4,3-c]pyridine ≥97%
7-Bromo-1H-pyrazolo[4,3-c]pyridine
Formula: C₆H₄BrN₃
MW: 198.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18256789
CAS Number: 1256821-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR