6-Bromo-2-methoxynicotinic acid ≥97%

Supplier: AOBChem USA

Warning

45553-1G 45553-25G 45553-500MG 45553-5G
76942-420EA 206.88 USD
76942-420 76942-422 76942-424 76942-426
6-Bromo-2-methoxynicotinic acid ≥97%
6-Bromo-2-methoxynicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD13188730
CAS Number: 1060806-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR