Ethyl 2-formylbenzoate ≥97%

Supplier: AOBChem USA

Warning

45539-1G 45539-25G 45539-500MG 45539-5G
76942-404EA 129.43 USD
76942-404 76942-406 76942-408 76942-410
Ethyl 2-formylbenzoate ≥97%
Ethyl 2-formylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MW: 178.185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09743805
CAS Number: 34046-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR