tert-Butyl (2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)biscarbamate ≥95%

Supplier: AOBChem USA

Warning

45007-1G 45007-25G 45007-500MG 45007-5G
76942-390EA 197.33 USD
76942-390 76942-392 76942-394 76942-396
tert-Butyl (2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)biscarbamate ≥95%
tert-Butyl (2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)biscarbamate
Formula: C₂₂H₃₂BF₂NO₆
MW: 455.3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30717696
CAS Number: 2377609-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR