Isopropyl 5-bromo-2-chloroisonicotinate ≥97%

Supplier: AOBChem USA

Warning

38735-25G 38735-5G
76942-368EA 275.84 USD
76942-368 76942-370
Isopropyl 5-bromo-2-chloroisonicotinate ≥97%
Isopropyl 5-bromo-2-chloroisonicotinate
Formula: C₉H₉BrClNO₂
MW: 278.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29932204
CAS Number: 1882662-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR