Ethyl-4-bromo-3-chlorobenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
Ethyl 4-bromo-3-chlorobenzoate
38722-1G
38722-25G
38722-5G
76942-354EA
84.88
USD
76942-354
76942-356
76942-358
Ethyl-4-bromo-3-chlorobenzoate ≥97%
Ethyl-4-bromo-3-chlorobenzoate
Formula:
C₉H₈BrClO₂ MW: 263.52 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD17779947 CAS Number: 120077-67-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...