Methyl 4-chloro-3-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

38690-1G 38690-25G 38690-500MG 38690-5G
76942-338EA 228.1 USD
76942-338 76942-340 76942-342 76942-344
Methyl 4-chloro-3-(methylthio)benzoate ≥97%
Methyl 4-chloro-3-(methylthio)benzoate
Formula: C₉H₉ClO₂S
MW: 216.685 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22489119
CAS Number: 166811-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR