6-(Benzyloxy)-2-fluoro-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

38647-1G 38647-25G 38647-500MG 38647-5G
76942-306EA 168.69 USD
76942-306 76942-308 76942-310 76942-312
6-(Benzyloxy)-2-fluoro-3-methylbenzoic acid ≥97%
6-(Benzyloxy)-2-fluoro-3-methylbenzoic acid
Formula: C₁₅H₁₃FO₃
Storage Temperature: Ambient
MDL Number: MFCD32707107
CAS Number: 2432848-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR