Ethyl 6-chloro-4-iodonicotinate ≥97%

Supplier: AOBChem USA

Warning

38551-1G 38551-500MG 38551-5G
76942-262EA 673.68 USD
76942-262 76942-264 76942-266
Ethyl 6-chloro-4-iodonicotinate ≥97%
Ethyl 6-chloro-4-iodonicotinate
Formula: C₈H₇ClINO₂
Storage Temperature: Refrigerator
MDL Number: MFCD31380039
CAS Number: 2383169-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR