5-Methyl-2-propoxybenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

38468-1G 38468-25G 38468-500MG 38468-5G
76942-238EA 195.21 USD
76942-238 76942-240 76942-242 76942-244
5-Methyl-2-propoxybenzaldehyde ≥97% research grade
5-Methyl-2-propoxybenzaldehyde
Formula: C₁₁H₁₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD06248399
CAS Number: 883531-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR