Ethyl 3-bromo-4-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

38401-1G 38401-25G 38401-500MG 38401-5G
76942-226EA 263.11 USD
76942-226 76942-228 76942-230 76942-232
Ethyl 3-bromo-4-methoxybenzoate ≥97%
Ethyl 3-bromo-4-methoxybenzoate
Formula: C₁₀H₁₁BrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD06204360
CAS Number: 460079-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR